missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novacyt - Microgen Bioproducts Rotascreen Latex
Microgen Bioproducts Rotascreen Latex - Rapid and simple latex agglutination test for the detection of Rotavirus antigen.
Brand: Novacyt - Microgen Bioproducts M80CE
This item is not returnable.
View return policy
Description
- A simple latex aggutination test for the detection of Adenovirus antigen in faecal specimens.
Specifications
ISO13485 ISO9001 | |
M80c Positive control - 1.0 mL | |
Non-waived | |
4 Bottles (2.5 mL, 2.5 mL, 1.5 mL, 50 mL) | |
0.97 | |
The Detection of Rotavirus Antigen in Faecal Specimens | |
50 tests | |
2 / test | |
Carton | |
M80a - Test reagent - 2.5 mL - Contains (rabbit) Rotavirus antibody-sensitised latex particles | |
0.97 | |
12 minutes |
2°C to 8°C | |
Yes | |
Rabbit Rotavirus antibody | |
Suspension of faecal specimen | |
Latex Agglutination Test | |
Laboratory Centrifuge, Stoppered or Screw-Capped Tubes | |
Polyethylene Dropper Bottles | |
Animal | |
4 Bottles (2.5 mL, 2.5 mL, 1.5 mL, 50 mL) | |
Rotavirus | |
Eukaryota |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novacyt - Microgen Bioproducts Rotascreen Latex > Rotascreen Latex
Spot an opportunity for improvement?Share a Content Correction