Learn More
Fisherbrand™ Rotator Drum
This Rotator Drum is designed for use with Fisherbrand™ Digital Cel-Gro Tissue Culture Rotator.
Brand:  Fisherbrand™ 88861241
 View more versions of this product
Product Code. 17487632
  • £239.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with Fisherbrand™ Digital Cel-Gro Tissue Culture Rotator
  • Various drum sizes available for rotating different sizes of tubes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
76 Tubes (26 mm dia.)
Rotating Drum