missing translation for 'onlineSavingsMsg'
Learn More

Scientific Industries Rotator Genie, Magnetic Platform

The Rotator Genie, Magnetic Platform is a rugged and versatile lab rotator featuring a magnetic platform designed to accept various magnetic clip plates (sold separately) to hold a variety of containers. Designed to fit in most incubators and cold rooms.

Brand:  Scientific Industries SI-2203

 View more versions of this product

Product Code. 15819645

This item is not returnable. View return policy

Description

Description

  • Holds a wide variety of tubes, flasks, microplates, bags, and odd-shaped vessels at any angle
  • Magnetic platform features unique clip plate system
  • May be used in cold rooms and incubators (Not for CO2 incubators)
  • Uniquely designed for a wide selection of motions adjustable from gentle rolling to forceful end over end mixing
  • Continuous-duty quality motor provides smooth, quiet operation
  • 2 Year Warranty
Specifications

Specifications

5 to 35 rpm
Magnetic Rotator
UK Plug
40.1 x 17.8 x 40.1 cm (15.8 x 7.0 x 15.8 in.)
5.9 kg
Rotator Genie
0.25 A, 50 Hz, 230 V
CE
29.2 x 31.8 cm (11.5 x 12.5 in.)
13 lb.
Rotating almost any container
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers