Learn More
BUCHI Rotavapor R-300 with Vertical Reflux, Electronic Lift
The Rotavapor R-300 meets the highest expectations of convenience and versatility in rotary evaporation. A modular design allows the R-300 to be extended to a fully integrated system, with a central interface regulating each component.
£4770.00 - £8475.00
Specifications
Weight (Metric) | 13.5 kg |
---|---|
Voltage | 100/240 V |
Speed | 10 to 280 rpm |
For Use With (Application) | Rotary Evaporation |
Cooling Surface | 1500 cm2 |
Product Code | Brand | Material (Coating) | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material (Coating) | Includes | Price | Quantity & Availability | ||||
15591110
|
BUCHI
11R300211S001 |
Uncoated | B-301 Heating Bath |
£4770.00
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
||||
15501120
|
BUCHI
11R300211S011 |
Uncoated | B-301 Heating Bath, Woulff Bottle |
£4955.00
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
||||
15531120
|
BUCHI
11R300211S041 |
Uncoated | B-301 Heating Bath, I-300 Interface, Valve Unit, and VacuBox |
£7045.00
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
||||
15511120
|
BUCHI
11R300211S021 |
Uncoated | B-301 Heating Bath, I-300 Interface, Woulff Bottle, and VacuBox |
£6665.00
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
||||
15541120
|
BUCHI
11R300211S051 |
Uncoated | B-301 Heating Bath, I-300 Pro Interface, Valve Unit, and VacuBox |
£8135.00
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
||||
15521120
|
BUCHI
11R300211S031 |
Uncoated | B-301 Heating Bath, I-300 Pro Interface, Woulff Bottle, and VacuBox |
£7755.00
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
||||
15521220
|
BUCHI
11R300251S001 |
Uncoated | B-305 Heating Bath |
£4980.00
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
||||
15531220
|
BUCHI
11R300251S011 |
Uncoated | B-305 Heating Bath, Woulff Bottle |
£5165.00
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
||||
15561220
|
BUCHI
11R300251S041 |
Uncoated | B-305 Heating Bath, I-300 Interface, Valve Unit, and VacuBox |
£7250.00
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
||||
15541220
|
BUCHI
11R300251S021 |
Uncoated | B-305 Heating Bath, I-300 Interface, Woulff Bottle, and VacuBox |
£6870.00
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
||||
15571220
|
BUCHI
11R300251S051 |
Uncoated | B-305 Heating Bath, I-300 Pro Interface, Valve Unit, and VacuBox |
£8340.00
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
||||
15551220
|
BUCHI
11R300251S031 |
Uncoated | B-305 Heating Bath, I-300 Pro Interface, Woulff Bottle, and VacuBox |
£7960.00
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
||||
15561120
|
BUCHI
11R300211S111 |
P+G Safety Coating | B-301 Heating Bath, Woulff Bottle |
£5080.00
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
||||
15591120
|
BUCHI
11R300211S141 |
P+G Safety Coating | B-301 Heating Bath, I-300 Interface, Valve Unit, and VacuBox |
£7180.00
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
||||
15571120
|
BUCHI
11R300211S121 |
P+G Safety Coating | B-301 Heating Bath, I-300 Interface, Woulff Bottle, and VacuBox |
£6800.00
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
||||
15501130
|
BUCHI
11R300211S151 |
P+G Safety Coating | B-301 Heating Bath, I-300 Pro Interface, Valve Unit, and VacuBox |
£8270.00
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
||||
15581120
|
BUCHI
11R300211S131 |
P+G Safety Coating | B-301 Heating Bath, I-300 Pro Interface, Woulff Bottle, and VacuBox |
£7875.00
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
||||
15591220
|
BUCHI
11R300251S111 |
P+G Safety Coating | B-305 Heating Bath, Woulff Bottle |
£5285.00
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
||||
15501230
|
BUCHI
11R300251S121 |
P+G Safety Coating | B-305 Heating Bath, I-300 Interface, Woulff Bottle, and VacuBox |
£7005.00
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
||||
15531230
|
BUCHI
11R300251S151 |
P+G Safety Coating | B-305 Heating Bath, I-300 Pro Interface, Valve Unit, and VacuBox |
£8475.00
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
||||
15511230
|
BUCHI
11R300251S131 |
P+G Safety Coating | B-305 Heating Bath, I-300 Pro Interface, Woulff Bottle, and VacuBox |
£8085.00
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
||||
Description
- A combination of convenience and versatility in rotary evaporation
- Modular design allows the R-300 to be extended to a fully integrated system
- Heating Bath B-305 is compatible with water or oil heating fluids
- Central interface compatibility to individually regulate each component
- User-friendly operation due to ergonomic electronic or manual lift options
Vertical Reflux condenser (with Shut-off Valve)
- For combining reflux reaction and distillation
- Connection for vapor temperature sensor
- Automatic distillation with Interface I-300/ I-300 Pro possible
- Not compatible with foam sensor
Smart Distillations
- Enables automated, unattended processes with optimized system parameters to take place
- Users can program their own SOPs into the system, then remotely monitor distillation processes
- BUCHI Cloud Services substantially maximize system uptimes with smart self-service tools and free updates
High Flexibility
- Modular design helps to meet individual requirements for rotary evaporation though high-level system flexibility
- A wide range of system accessories, glassware, and mobile monitoring options are available
- Users are capable of customizing their specific distillation process to fit any desired application
- Seven different glass assemblies that are compatible with various solvents and sample properties
Maximum Convenience
- The R-300 can be easily operated with features such as one-touch interface commands, and fast flask handling with Combi-Clip
- Combi-Clip with snap lock mechanism allows one-handed connection of evaporating flasks
- Options to manually adjust system parameters with intuitively placed buttons
- Immediate adjustment of heating temperature and rotation speed in the case of an urgent or unexpected event
- Exclusive possibility to personalize glassware with a customized permanent print
Specifications
13.5 kg | |
10 to 280 rpm | |
1500 cm2 | |
Vertical Reflux | |
10° to 50° | |
Rotary Evaporator | |
Rotavapor R-300 |
100/240 V | |
Rotary Evaporation | |
100 W | |
IP 21 | |
CE/CSA Approved | |
50/60 Hz | |
29/32 |