Learn More
BUCHI Rotavapor Vapor Duct with Combi-Clip
Vapor duct with Combi-Clip for use with connecting the evaporating flask to the Rotavapor.
Brand:  BUCHI 11062186
 View more versions of this product
Product Code. 15475875
  • £196.00 / Each
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Combi-Clip is included for fastening the evaporating flask
Specifications
Specifications
Borosilicate Glass 3.3
Glass Assembly V, C, S, E, CR, BY, HP
Combi-Clip
Vapor Duct
Rotavapor R-300
29/32