Learn More
IKA™ ROTAVISC Vane Spindle
Vane spindle designed for use with IKA™ ROTAVISC Viscometers.
Brand: IKA™ 0025006856
This item is not returnable.
View return policy
Description
- Quick coupling
- Hook hitch
- M2.5 left-handed thread
Specifications
Vane Spindle | |
IKA™ ROTAVISC Viscometer | |
VAN-SP-4 Vane Spindle | |
AISI 316L |
Product Content Correction
Product Title
IKA™ ROTAVISC Vane Spindle > Vane Viscometer Spindle; Description: VAN-SP-4 Vane Spindle
Spot an opportunity for improvement?Share a Content Correction