Learn More
IKA™ ROTAVISC Vane Spindle
Vane spindle designed for use with IKA™ ROTAVISC Viscometers.
Brand: IKA™ 0025008376
This item is not returnable.
View return policy
Description
- Quick coupling
- Hook hitch
- M2.5 left-handed thread
Specifications
Vane Spindle | |
IKA™ ROTAVISC Viscometer | |
VAN-SP-5 Vane Spindle | |
AISI 316L |