Learn More
IKA™ ROTAVISC Vane Spindle
Vane spindle designed for use with IKA™ ROTAVISC Viscometers.
Brand:  IKA™ 0025006854
 View more versions of this product
Product Code. 17287549
  • £281.00 / Each
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Quick coupling
  • Hook hitch
  • M2.5 left-handed thread
Specifications
Specifications
Vane Spindle
IKA™ ROTAVISC Viscometer
VAN-SP-2 Vane Spindle
AISI 316L