missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rotenone, ≥97%, MP Biomedicals™
Click to view available options
Quantity:
1 g
5 g
Specifications
Color | Beige to White |
Assay Percent Range | ≥97% |
Quantity | 1 g |
Formula Weight | 394.4 |
Physical Form | Powder |
Chemical Name or Material | Rotenone |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H301-Toxic if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : rotenone
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'