Learn More
Eppendorf™ Rotor Adapters for Centrifuge 5427 R
For 1.5/2mL tubes and cryotubes
Brand:  Eppendorf™ 5820769009
 View more versions of this product
Product Code. 13226619
  • £38.10 / Pack of 4
Explore more special offers
Description
Description
  • Rotors FA-45-12-17, FA-45-16-17 and FA-45-20-17
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rotor Adapter
1 Cryotube
Set of 4
Cryotube
Rotors FA-45-12-17, FA-45-16-17 and FA-45-20-17