Learn More
Fisherbrand™ Rotor Knob
Remove old knob with counter-clockwise turns. Fisherbrand™ Rotor Knob will engage with clockwise turns.
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 0.10000kg
Description
For use with Mini-CentrifugeServices
Five Years
Specifications
Rotor Knob | |
Fisherbrand Mini-centrifuge |