Learn More
Eppendorf™ Rotor Lids
Product Code. p-8004769 Shop All Eppendorf Products
Click to view available options
For Use With (Equipment):
A-2-MTP
Centrifuge 5427R, FA-45-24-11-Kit
Centrifuge 5427R, FA-45-30-11
F-34-6-38
F-45-17-17-Cyro
F-45-30-11
F-45-48-11
F-45-64-5-PCR
FA-45-16-17
FA-45-24-11
FA-45-24-11-HS
FA-45-24-11-Kit
FA-45-30-11
FA-45-48-11
S-24-11-AT
Material:
Aluminum
Aluminum, Coated
Polypropylene
Description:
Aerosol-tight
Aerosol-tight, QuickLock
Coated
Non-aerosol-tight
Wind shield upper shell
Unit Size:
Each
Product Code. 12334948
Brand: Eppendorf™ 5427725008
Designed for use with Eppendorf Rotors
  • Replacement rotor lids
  • Aerosol-tight and aluminum lids available
  • Available in a variety of sizes
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor Lid
Material Aluminum
For Use With (Equipment) A-2-MTP
Description Wind shield upper shell