Learn More
Eppendorf™ Rotor for Centrifuges 5910 Ri and 5920 R
Eppendorf Rotor for Centrifuge 5910 Ri and 5920 R-precision in refrigerated large-capacity centrifugation. Tailor your experiments with varied capacities and speeds, buckets and adaptors for efficient sample processing. Ideal for high-throughput applications, ensuring accuracy and reproducibility.
Brand: Eppendorf™ 5895121004
1282.00 GBP valid until 2024-12-31
Use promo code "23292" to get your promotional price.
Additional Details : Weight : 5.00000kg
Descripción
- Broad range of adapters enables high versatility for tubes and flasks from 0.2mL to 750mL and plates
- Mixed loading with 2 plate buckets and 2 round tube buckets allows for increased versatility
- Aerosol-tight QuickLock caps for safe centrifugation of hazardous samples optionally available
- Buckets, caps and adapters are autoclavable
Especificaciones
Rotor S-4x750, without buckets |