Learn More
Eppendorf™ Rotor for Centrifuge 5920 R
Eppendorf Rotor for Centrifuge 5920 R-precision in refrigerated large-capacity centrifugation. Tailor your experiments with varied capacities and speeds. Accessorize with versatile buckets and adaptors for efficient sample processing. Ideal for high-throughput applications, ensuring accuracy and reproducibility.
Brand: Eppendorf™ 5895190006
Additional Details : Weight : 9.45000kg
Description
- Broad range of adapters enables high versatility for tubes and flasks from 0.2mL to 750mL and plates
- Universal bucket design accommodates both plates and tubes
- Aerosol-tight QuickLock caps for safe centrifugation of hazardous samples optionally available
- Buckets, caps and adapters are autoclavable
Specifications
Rotor S-4x Universal-Large | |
4,402 x g | |
Eppendorf 5920R | |
Autoclavable |
4 x 750 mL | |
4,250 rpm | |
without buckets |