missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Rotor for Centrifuge 5910 Ri

Eppendorf Rotor for Centrifuge 5910 Ri-precision in refrigerated centrifugation. Tailor your experiments with varied capacities and speeds, adaptable buckets and adaptors for efficient sample processing. Ideal for temperature-sensitive applications, ensuring accuracy and reproducibility.

Brand:  Eppendorf™ 5895181007

1177.00 GBP valid until 2024-12-31
Use promo code "23292" to get your promotional price.



Additional Details : Weight : 6.00000kg

 View more versions of this product

Product Code. 15833212

  • £1240.00 / Each
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • High-speed medium capacity swing-bucket rotor
  • Broad range of adapters enables high versatility for tubes and bottles
  • Aerosol-tight QuickLock caps for safe centrifugation of hazardous samples optionally available
  • Buckets, caps and adapters are autoclavable
  • Requires buckets to accommodate tubes and bottles from 0.2 to 250 mL
Specifications

Specifications

0.2 to 250 mL
5910 R Centrifuge
4
5,100 rpm
Without buckets
Aerosol-tight caps available
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers