Learn More
Grant Instruments™ Rotor for PCV Series Compact Benchtop Centrifuges
Fits PCV-2400 Combi-Spin and PVC6400 Multi-spin benchtop centrifuges. Grant Intstruments™ Rotor for PCV Series Compact Benchtop Centrifuges provide versatility with interchangeable rotors for various microtube sizes.
£38.16 - £47.00
Products 3
Product Code Brand No. of Places Price Quantity & Availability  
Product Code Brand No. of Places Price Quantity & Availability  
11481028
View Documents Promotion Details
Grant Instruments™
PSR-16
16 (2 microtube strips)
£47.00
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
11451028
View Documents Promotion Details
Grant Instruments™
PR2-05
16
£38.16
Each
View alternative products
Save Up to 
»
Estimated Shipment: 20-08-2024
to see stock.
11471028
View Documents Promotion Details
Grant Instruments™
PR2-05-02
18
£38.16
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Description
Description
  • PR2-05: Interchangeable centrifuge rotor for 8 x 1.5/2.0mL plus 8 x 0.5mL microtubes (optional)
  • PR2-05-02 : Interchangeable centrifuge rotor for 6 x 1.5/2.0mL plus 6 x 0.2mL microtubes (optional)
  • PSR-16: Interchangeable centrifuge rotor for 2 x 8-well 0.2mL microtube strips (optional)