Learn More
SVL™ Rotulex™ Spherical Joints
Offers a grease free spherical joint
Brand:  SVL™ RM41
Code : NEW
Additional Details : Weight : 0.56000kg
 View more versions of this product
Product Code. 15507960
  • £1405.00 / Pack of 30
Estimated Shipment: 03-10-2024
to see stock.
Add to basket
Description
Description
  • All glass and PTFE pathway is highly resistant to chemical attack
  • Can be used at temperatures up to 200°C
  • System comprises male ball joint with PTFE covered O-ring which fits into female cup joint
  • Joint can be retained together with spring loaded metal clip
  • Component parts RM, RF, RO and RC should be ordered separately
Specifications
Specifications
Male joint, PTFE covered O-ring, spring loaded Metal clip
Glass, PTFE
41