Learn More
SVL™ Rotulex™ Spherical Joints
Offers a grease free spherical joint
Brand:  SVL™ RM64
Code : NEW
Additional Details : Weight : 0.75000kg
 View more versions of this product
Product Code. 15517960
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • All glass and PTFE pathway is highly resistant to chemical attack
  • Can be used at temperatures up to 200°C
  • System comprises male ball joint with PTFE covered O-ring which fits into female cup joint
  • Joint can be retained together with spring loaded metal clip
  • Component parts RM, RF, RO and RC should be ordered separately
Specifications
Specifications
Male joint
Glass, PTFE
5Pack
64