Learn More
IKA™ Round Bottom Flask Carrier
Compatible with several IKA stirrers.
Brand:  IKA™ 0025000833
 View more versions of this product
Product Code. 15254037
  • £272.00 / Each
Estimated Shipment: 19-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Round/hollow cylindrical bottom
  • Aluminum anodized
Specifications
Specifications
1000 mL
Round Bottom
Flask Carrier
Without handle
180°C