missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKA™ Round Bottom Flask Inlay
Round bottom flask inlay constructed of anodized aluminum
£88.85 - £161.00
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15224007
|
IKA™
0025000637 |
25 mL |
£88.85
Each |
In Stock
Log in to see stock. |
|||||
15178449
|
IKA™
0025000638 |
50 mL |
£96.95
Each |
In Stock
Log in to see stock. |
|||||
15213305
|
IKA™
0025000636 |
10 mL |
£88.85
Each |
In Stock
Log in to see stock. |
|||||
15214037
|
IKA™
0025000641 |
100 mL |
£96.95
Each |
Estimated Shipment: 28-02-2025
Log in to see stock. |
|||||
15284037
|
IKA™
0025000644 |
500 mL |
£161.00
Each |
In Stock
Log in to see stock. |
|||||
Description
- Round bottom
- Anodized aluminum construction
- Temperature resistant
Specifications
Round Bottom | |
180°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title