Learn More
Lenz Laborglasinstrumente™ Test Tubes with NS Joint, without Graduation
Ideal for routine laboratory analysis and procedures. Lenz Laborglasinstrumente™ Test Tubes with NS Joint, without Graduation are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 05700007
Additional Details : Weight : 0.15300kg
 View more versions of this product
Product Code. 10396412
  • £46.00 / Pack of 10
Estimated Shipment: 28-08-2024
to see stock.
Add to basket
Description
Description
  • With round bottom and NS socket
  • Available without stopper, with PE stopper, or with hexagonal hollow glass stopper
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
Glass Tube
Neck Size: 14/23; Without Stopper