missing translation for 'onlineSavingsMsg'
Learn More

SCAT Round Canisters

Catalog Number 10801511
31.23 GBP valid until 2024-12-31
Use promo code "23290" to get your promotional price.


Click to view available options
Capacity (Metric):
2 L
4 L
5 L
10 L
25 L
50 L
60 L
Closure Size:
53B
83B
S65

SCAT round canisters are ideal for collecting and disposing of smaller quantities of solvents due to their compact dimensions. Several SCAT round canisters can be easily stationed at the HPLC system and equipped with SCAT SafeyWasteCaps or the SCAT Universal Waste Hub 'Jan'.

  • Made of either robust HDPE or PP and therefore resistant to all solvents commonly used in the laboratory
  • Available in the common threads S65, S70/71 and B83
  • Filling capacities range from very compact 2 L to 4 L, 10 L, 25 L, 50 L and 60 L for the collection of voluminous solvents

Many individual features such as carrying handles, handles or measuring scales, which ensure safe handling of the hazardous substances
SCAT round canisters are a perfect alternative or supplement to conventional SCAT canisters to provide physical separation of solvents and prevent mixing of hazardous liquids.

Capacity (English) 0.5 gal.
Capacity (Metric) 2 L
Closure Size 53B
Color White (Translucent)
Description Round canister 2 L, B53, PE-HD, dimensions (W x H x D): 119 x 260 x 119 mm
Dimensions (L x W x H) 119 x 260 x 119 mm
Disposable Reusable
Material HDPE
Mouth Style B53 Thread
Product Type Canister
Shape Round
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'