Learn More
W.S. TYLER™ Round Sieve Brush
W.S. Tyler round sieve brushes are ideal for cleaning fine and coarse mesh sieves, 120 mesh and finer. Proper maintenance and cleaning of sieves prevents damage to sieve openings, keeping them clean and free of particles, and ensures the longest possible in-spec service life and accurate results.
Brand: W.S. TYLER™ 59986-96
Code : A0
Additional Details : Weight : 0.80000kg
Description
- For 120 mesh sieves, and finer
- Keeps sieves clean for better separation
- Will not damage or hinder sieve performance
- Nylon bristles
Specifications
Nylon Bristle, Wooden Handle | |
Sieve Cleaning | |
Clean 120 Mesh and Finer |
Sieve Cleaning Brush | |
Test Sieves |