missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPL27A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564112
This item is not returnable.
View return policy
Description
Immunogen sequence: PSRLRKTRKL RGHVSHGHGR IGKHRKH Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Rpl27a gene ontology annotations related to this gene include translation.
Specifications
| RPL27A | |
| Polyclonal | |
| Unconjugated | |
| RPL27A | |
| 60S ribosomal protein L27a; 60S ribosomal protein L27-A; 60S ribosomal protein L27a-like protein; fa93g12; FLJ43464; hypothetical protein LOC336726; L27'; L27A; L29; Large ribosomal subunit protein eL27-A; Large ribosomal subunit protein uL15; MGC10850; MGC17878; MGC87238; ORF; ORF protein; ribosomal 60S subunit protein L27A; ribosomal protein L27A; ribosomal protein L27a-like; ribosomal protein L29 homolog; RPL27; RPL27A; wu:fa93g12; YHR010W; zgc:77235 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6157 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P46776 | |
| RPL27A | |
| Recombinant protein corresponding to Human RPL27A. Recombinant protein control fragment (Product #RP-103507). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction