Learn More
RPL3 Mouse anti-Human, CoraLite 488, Clone: 2G1B8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-66130-100UL
Product Code. 16810736
  • £375.00 / 100µL
Explore more special offers
Description
Description
The RPL3 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human RPL3. This antibody recognizes human, mouse, rat antigen. The RPL3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
Specifications
RPL3
Monoclonal
CoraLite 488
100 μL
6122
IgG1
Immunofluorescence
2G1B8
Mouse
Primary
Human, Rat, Mouse
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
RPL3 Mouse anti-Human, CoraLite 488, Clone: 2G1B8, Proteintech > 100 μL