missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL35 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14826-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L29P family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.Specifications
RPL35 | |
Polyclonal | |
Unconjugated | |
RPL35 | |
60S ribosomal protein L35, ribosomal protein L35, RPL35 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11224, 296709, 66489 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.46 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P17078, P42766, Q6ZWV7 | |
Rpl35 | |
RPL35 Fusion Protein Ag6615 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |