missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL37A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14660-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit.Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L37AE family of ribosomal proteins. It is located in the cytoplasm. The protein contains a C4-type zinc finger-like domain. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.Specifications
RPL37A | |
Polyclonal | |
Unconjugated | |
RPL37A | |
60S ribosomal protein L37a, ribosomal protein L37a, RPL37A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100361520, 19981, 6168 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
D4AAZ6, P61513, P61514 | |
RPL37A | |
RPL37A Fusion Protein Ag6330 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |