Learn More
Corning™ RPMI 1640, 1X
Glutagro™ supplement provides greatly improved stability over the L-glutamine media supplement
Brand:  Corning™ 17-105-CV
Code : NEW
Additional Details : Weight : 3.76000kg
 View more versions of this product
Product Code. 15363561
  • £159.93 / Pack of 6
In Stock
to see stock.
Description
Description
Named for the site at which it was developed, the Roswell Park Memorial Institute, RPMI 1640 is a modification of McCoy's 5A containing an increased level of inositol. Cell types for which this medium is appropriate include mammalian myeloma and hybridoma cells such as mouse hybridomas, human leukocytes, and B and T lymphocytes.
  • Can be used for suspension cultures or monolayer human leukemic cells
  • This formulation is supplemented with glutaGRO™ in place of L-glutamine for improved stability
Specifications
Specifications
2° to 8°C
RPMI 1640 w/o L-glut and ph.red
<0.25EU/mL
To assess growth promotion and cytotoxicity characteristics
6/Pk.
1 X
No Glutamine, No Phenol Red
L-Cystine/L-Methionine, Sodium Bicarbonate
7 to 7.4