missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ RPMI 1640 Medium
RPMI 1640 Medium
Brand: Gibco™ 21875059
23.41 GBP valid until 2024-12-31
Use promo code "21941" to get your promotional price.
This item is not returnable.
View return policy
Description
Roswell Park Memorial Institute (RPMI) 1640 medium was originally developed to culture human leukemic cells in suspension and as a monolayer. RPMI 1640 has since been found suitable for a variety of mammalian cells including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas. We offer a variety of Gibco™ RPMI 1640 modifications for a range of cell culture applications.Gibco™ RPMI 1640 is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 contains biotin, vitamin B12, and PABA which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law. Customers using Gibco™ RPMI 1640 in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).
cGMP Manufacturing and Quality System
Gibco™ RPMI 1640 is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer an identical Gibco™ RPMI 1640 product made in our Grand Island facility (11875-127). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
RPMI 1640 contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). RPMI 1640 uses a sodium bicarbonate buffer system (2.0 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Specifications
ISO 13485 standards, FDA | |
RPMI 1640 Medium, 1 X Concentration, 1000mL, Liquid Form, With L-glutamine, Phenol Red, 7 to 7.4 pH, 12 Months Shelf Life, 260 to 310mOsm/kg Osmolality, 2 to 8°C Storage Conditions, Soluble in Water, Used for Mammalian Cell Culture | |
Liquid | |
Glutamine, Phenol Red | |
Mammalian cell culture | |
1000 mL |
2 to 8°C | |
1 X | |
RPMI 1640 Medium | |
No HEPES, No Sodium Pyruvate | |
7 to 7.4 | |
For in-vitro diagnostic use |