missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ RPMI 1640 Medium, GlutaMAX™ Supplement, HEPES
GREENER_CHOICE

Product Code. p-7076214
Click to view available options
Quantity:
10 x 500mL
100 mL
20 x 100mL
500mL
Unit Size:
100mL
5000mL
500mL
Pack of 20
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11534526

Brand: Gibco™ 72400013

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

RPMI 1640 Medium, GlutaMAX™ Supplement, HEPES

Roswell Park Memorial Institute (RPMI) 1640 medium was originally developed to culture human leukemic cells in suspension and as a monolayer. RPMI 1640 has since been found suitable for a variety of mammalian cells including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes and carcinomas. We offer a variety of Gibco™ RPMI 1640 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

Gibco™ RPMI 1640 is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 contains biotin, vitamin B12 and PABA which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations. Gibco™ RPMI 1640 with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law. Customers using Gibco™ RPMI 1640 in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).

Dual-Site cGMP Manufacturing
Gibco™ RPMI 1640 is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ RPMI 1640 product made in our Grand Island facility (72400-047). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

RPMI 1640 contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). RPMI 1640 uses a sodium bicarbonate buffer system (2.0 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Product Type Roswell Park Memorial Institute (RPMI) 1640 Medium
With Additives GlutaMAX, Phenol Red, HEPES
Without Additives No Sodium Pyruvate
For Use With (Application) Mammalian cell culture
Quantity 100 mL
Certifications/Compliance ISO 13485 standards, FDA
Content And Storage 2 to 8°C
Description Roswell Park Memorial Institute (RPMI) 1640 Medium, 100mL, Liquid, HEPES Buffer, Phenol Red Indicator, With Glutamax™ I, Phenol Red, HEPES, Standard Serum Supplementation, For Mammalian Cell Culture
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.