missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ RPMI 1640 Medium, no glutamine

RPMI 1640 Medium, no glutamine
£16.89 - £154.00
Specifications
Concentration | 1 X |
---|---|
Form | Liquid |
Product Type | RPMI 1640 Medium |
With Additives | Phenol Red |
Without Additives | No Glutamine, No HEPES, No Sodium Pyruvate |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11534446
![]() |
Gibco™
31870025 |
500 mL |
£16.89
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11544446
![]() |
Gibco™
31870074 |
10 x 500 mL |
£154.00
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Roswell Park Memorial Institute (RPMI) 1640 medium was originally developed to culture human leukemic cells in suspension and as a monolayer. RPMI 1640 has since been found suitable for a variety of mammalian cells including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas. We offer a variety of Gibco™ RPMI 1640 modifications for a range of cell culture applications.Gibco™ RPMI 1640 is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 contains biotin, vitamin B12, and PABA which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations.
Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.
cGMP Manufacturing and Quality System
Gibco™ RPMI 1640 is manufactured at a cGMP compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.
RPMI 1640 contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). RPMI 1640 uses a sodium bicarbonate buffer system (2.0 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Specifications
1 X | |
RPMI 1640 Medium | |
No Glutamine, No HEPES, No Sodium Pyruvate | |
ISO 13485 standards, FDA | |
For in-vitro diagnostic use | |
RPMI 1640 Medium, 500mL, 1 X Concentration, Liquid Form, With Phenol Red, 7 to 7.4 pH, 12 Months Shelf Life, 260 to 310mOsm/kg Osmolality, 2 to 8°C Storage Conditions, Used for Mammalian Cell Culture |
Liquid | |
Phenol Red | |
Mammalian cell culture | |
7 to 7.4 | |
2 to 8°C |
Safety and Handling
missing translation for 'shelfLife' : 12 Months
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ RPMI 1640 Medium, no glutamine