missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ RPMI 1640 Medium, no glutamine
GREENER_CHOICE

Product Code. 11524446
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11524446

Brand: Gibco™ 31870017

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Roswell Park Memorial Institute (RPMI) 1640 medium was originally developed to culture human leukemic cells in suspension and as a monolayer. RPMI 1640 has since been found suitable for a variety of mammalian cells including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes and carcinomas.

Roswell Park Memorial Institute (RPMI) 1640 medium was originally developed to culture human leukemic cells in suspension and as a monolayer. RPMI 1640 has since been found suitable for a variety of mammalian cells including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes and carcinomas. We offer a variety of Gibco™ RPMI 1640 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This RPMI 1640 is modified as follows:

With: Phenol Red

Without: L-glutamine, HEPES

The complete formulation is available.

Gibco™ RPMI 1640 is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 contains biotin, vitamin B12 and PABA which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations.

Customers using Gibco™ RPMI 1640 in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).

RPMI 1640 contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). RPMI 1640 uses a sodium bicarbonate buffer system (2.0 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Mammalian Cells
Concentration 1 X
Form Liquid
Product Type RPMI 1640
Serum Level Standard Serum Supplementation
Sterility Sterile-filtered
With Additives Phenol Red
Without Additives No Glutamine, No HEPES, No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco
Quantity 100 mL
Shipping Condition Room Temperature
Content And Storage Store in refrigerator (2–8°C).
Culture Type Mammalian Cell Culture
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.