missing translation for 'onlineSavingsMsg'
Learn More

Ribophorin II Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA552540

Product Code. 15740491

  • £399.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Immunogen sequence: SISNETKDLL LAAVSEDSSV TQIYHAVAAL SGFGLPLASQ EALSALTARL SKEETVLATV QALQTASHLS QQADLRSIVE EIEDLVARLD ELGGVYLQFE EGLETTALFV AATYKLMDHV GTE Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.

RNP2 a type I integral membrane protein found only in the rough endoplasmic reticulum. The encoded protein is part of an N-oligosaccharyl transferase complex that links high mannose oligosaccharides to asparagine residues found in the Asn-X-Ser/Thr consensus motif of nascent polypeptide chains. This protein is similar in sequence to the yeast oligosaccharyl transferase subunit SWP1.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ribophorin II
Polyclonal
Unconjugated
RPN2
1300012C06Rik; AV261018; Dolichyl-diphosphooligosaccharide--protein glycosyltransferase 63 kDa subunit; dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit 2; hypothetical protein LOC534231; oligosaccharyltransferase complex subunit (non-catalytic); RIBIIR; ribophorin 2; ribophorin II; Ribophorin-2; RPN2; Rpn-2; RPNII; RPN-II; SWP1
Rabbit
Antigen affinity chromatography
RUO
6185
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry, Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P04844
RPN2
Recombinant protein corresponding to Human RPN2
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only