Learn More
RPS3 Mouse anti-Human, CoraLite 488, Clone: 2G7H4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-66046-100UL
Product Code. 16820706
  • £375.00 / 100µL
Explore more special offers
Description
Description
The RPS3 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human RPS3. This antibody recognizes human, mouse, rat antigen. The RPS3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
Specifications
RPS3
Monoclonal
CoraLite 488
100 μL
6188
IgG2a
Immunofluorescence
2G7H4
Mouse
Primary
Human, Rat, Mouse
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
RPS3 Mouse anti-Human, CoraLite 488, Clone: 2G7H4, Proteintech > 100 μL