missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPS6KA3 (RSK2) Recombinant Human Protein, Inactive
RPS6KA3 (RSK2) is a serine/threonine kinase.
Brand: Invitrogen™ PV3289
Code : NEW
Additional Details : Weight : 0.00900kg
Description
RPS6KA3 (RSK2) modulates many cellular processes including transcription, adhesion, growth, survival, and proliferation.
Specifications
NP_004577 | |
50mM Tris, pH 7.5, 150mM NaCl, 1mM EDTA, 0.05% Triton X-100, 5mM DTT, 50% glycerol | |
86.3kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length, Recombinant, Inactive | |
ribosomal protein S6 kinase, 90kDa, polypeptide 3 | |
Human | |
His Tag | |
Kinases & Inhibitors |
Cell Analysis, Cell Signaling, Protein Biology, Target and Lead Identification and Validation, Cell Signaling, Kinase Assay, Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Cell Proliferation and Function | |
6197 | |
100μg | |
RUO - research use only | |
RPS6KA3 (RSK2) | |
RSK (RPS6K) Kinase Family | |
Recombinant | |
Baculovirus | |
Serine/Threonine Kinases |