missing translation for 'onlineSavingsMsg'
Learn More

(RS)-AMPA, Tocris Bioscience™

Product Code. 30001056
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective AMPA agonist

Prototypical and defining agonist for the AMPA subgroup of ionotropic glutamate receptors.

Active Enantiomer, Inactive Enantiomer and Hydrobromide Salt also available.

Chemical Identifiers

CAS 74341-63-2
Molecular Formula C7H10N2O4
Molecular Weight (g/mol) 186.167
InChI Key UUDAMDVQRQNNHZ-UHFFFAOYSA-N
Synonym ampa, r,s-ampa, ampa pharmaceutical, alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, rs-ampa, +/--ampa, chembl13378, d,l-alpha-amino-3-hydroxy-5-methylisoxazole-4-propionic acid, r,s-alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, 4-isoxazolepropanoic acid, alpha-amino-2,3-dihydro-5-methyl-3-oxo
PubChem CID 1221
ChEBI CHEBI:34018
IUPAC Name 2-amino-3-(5-methyl-3-oxo-1,2-oxazol-4-yl)propanoic acid
SMILES CC1=C(C(=O)NO1)CC(C(=O)O)N

Specifications

Quantity 10 mg
Formula Weight 186.17
Percent Purity >99%
Chemical Name or Material (RS)-AMPA
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'