Learn More
Tocris Bioscience™ (RS)-(Tetrazol-5-yl)glycine
Highly potent NMDA agonist
Brand: Tocris Bioscience™ 0312/50
This item is not returnable.
View return policy
Description
(RS)-(Tetrazol-5-yl)glycine is a potent NMDA receptor agonist, approximately 20 times more active than NMDA.
Specifications
138199-51-6 | |
50 mg | |
NMDA Receptor Agonists | |
Observed MW: 143.1 |
>98% | |
Soluble to 50 mM in water | |
C3H5N5O2 | |
98% |
Product Content Correction
Product Title
Tocris Bioscience™ (RS)-(Tetrazol-5-yl)glycine > 50 mg