Learn More
Invitrogen™ RSV Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA14104
Description
PA1-4104 detects human and bovine respiratory syncytial virus. PA1-4104 has been successfully used in Western blot procedures. The PA1-4104 immunogen is a synthetic peptide corresponding to residues F(483) P S D E F(488) C of human and bovine RSV. This sequence is completely conserved among all known species.
Human respiratory syncytial virus (RSV) is a member of the Paramyxoviridae family, subfamily Pneumovirinae, that causes common respiratory tract infections such as measles and mumps. Its genome consists of single stranded, negative sense RNA that encodes three envelope glycoproteins, a small hydrophobic (SH) protein of unknown function, a major glycoprotein (G) known as the attachment protein, and a fusion (F) protein. Its name comes from the fact that F proteins on the surface of the virus cause the membranes on nearby cells to merge, forming syncytia. While RSV typically produces mild symptoms in adults, it is one of the leading causes of lower respiratory tract infections and hospital visits during infancy and childhood.
Specifications
| RSV | |
| Polyclonal | |
| Unconjugated | |
| human respiratory syncytial virus; RSV | |
| Synthetic peptide corresponding to residues F(483) P S D E F(488) C of human and bovine RSV. | |
| 100 μg | |
| Primary | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.