missing translation for 'onlineSavingsMsg'
Learn More

RT1.D Monoclonal Antibody (OX-17), Biotin, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA517435

Product Code. 15338340

  • £221.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

anti-rat RT1.D monoclonal antibody recognizes a monomorphic determinant on the α chain of the rat Ia antigen and appears to be the rat homologue of mouse Ia-E. It recognizes the rat Ia product present on B, but not T cells from lymph node or thoracic duct lymph. It does not bind to thymocytes or erythrocytes. The antibody does not cross-react with rat Ia-A or mouse Ia-E antigen, but rabbit antibody raised against the antibody affinity column-purified MRC OX-17 antigen cross-reacted on tissues of mice expressing Ia-E mouse antigen but not on those mouse strains that were Ia-E antigen negative.

RT1.D Monoclonal antibody specifically detects RT1.D in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RT1.D
Monoclonal
0.1 mg/mL
PBS with 4-5mg/mL BSA and 0.02% sodium azide
P18211
RT1-Db1
Rat spleen membrane glycoproteins depleted of Ia-A antigens.
100 μg
Primary
Rat
Antibody
IgG1
Flow Cytometry, Immunohistochemistry (Frozen)
OX-17
Biotin
RT1-Db1
Db1; MHC class II RT1D beta1 chain antigen; rano class II histocompatibility antigen, D-1 beta chain; rat class I D; RT1 class II histocompatibility antigen, D-1 beta chain; RT1 class II, locus Db1; RT1-D beta; RT1-Db; RT1-Db1; RT1-Db1n
Mouse
Protein G
RUO
294270
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.