Learn More
RT1B Mouse anti-Rat, FITC, Clone: OX-6, BD
Mouse Monoclonal Antibody
£171.00
Specifications
Antigen | RT1B |
---|---|
Clone | OX-6 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15801619
|
BD Biosciences
562067 |
50 μg | |||||||
Description
The OX-6 antibody reacts with non-polymorphic determinants of the Rat MHC class II antigen, I-A equivalent. RT1B is found on peripheral B lymphocytes, thymic cortical epithelial and medullary reticular cells, small intestinal villus epithelium, epidermal Langerhans cells, dendritic cells, some tissue macrophage populations, peritoneal mast cells, and a subset of thymocytes, but not on peripheral T cells, erythrocytes, or microglia. The OX-6 mAb cross-reacts with mouse I-A[k] and I-A[s] alloantigens and with a major subset of splenocytes from NOD (I-A[g7]) mice.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
RT1B | |
0.5mg/mL | |
Monoclonal | |
Mouse | |
Mouse, Rat | |
Ia-like Glycoproteins from Wistar Thymocytes | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
OX-6 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified |
For Research Use Only.