missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RU 28318, potassium salt, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and selective antagonist for the mineralocorticoid receptor (MR). Inhibits aldosterone production and secretion, and selectively decreases ex-vivo MR binding in the hippocampus of adrenalectomised rats. Also decreases blood pressure in female rats following central administration in vivo.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 440.66 |
Percent Purity | >98% |
Chemical Name or Material | RU 28318, potassium salt |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'