Learn More
Abeba™ Rubber 5200 Shoes
ESD dissipative shoe is acid and base resistant.
Brand: Abeba™ 5200/43
51.48 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Additional Details : Weight : 1.00000kg
Description
- CE EN ISO 20347:2012 OB SRC
- Smooth leather white
- PU midsole
- Slip resistance special rubber outsole sole
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap foldable, adjustable
- Leather top layer
Specifications
CE EN ISO 20347:2012 OB SRC | |
Leather | |
Slip-resistant | |
5200 |
White | |
43 | |
Rubber |