Learn More
Abeba™ Rubber 9310 Shoes
Acid and base resistant. Rubber sole provides high slip resistance.
Brand:  Abeba™ 9310/38
 View more versions of this product
Product Code. 15663238
  • £58.80 / 1 pair
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather, perforated
  • Black
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Sanitized treated
  • Antistatic
  • Heel strap foldable, adjustable
  • Leather insole
Specifications
Specifications
CE EN ISO 20347:2012 OB A, FO SRC/li
Leather
Slip-resistant
9310
Black
38
Rubber