Learn More
Fisherbrand™ Rubber Cone Filter Crucible Adapters
Fisherbrand™ Rubber Cone Filter Crucible Adapters facilitate safe and secure placement of filter crucibles into a funnel.
Brand:  Fisherbrand™ CWF-820-050R
 View more versions of this product
Product Code. 12318855
  • £43.45 / Pack of 10
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Available in a range of sizes
  • Provided in packs of 10
Specifications
Specifications
40mm I.D. large end; 25mm I.D. small end
45mm
Cone
Filter crucibles
Rubber
10Pack