missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint Gobain Life Sciences Rubber Gas Sample Bag
Use these bags for gas sample analysis. Saint Gobain Life Sciences Rubber Gas Sample Bags feature a cylindrical shape and an ebonite tube and tap.
£50.20 - £169.00
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11754498
|
Saint Gobain Life Sciences
5607076 |
5 L | N/A | ||||||
11744498
|
Saint Gobain Life Sciences
5607074 |
2 L | N/A | ||||||
11724498
|
Saint Gobain Life Sciences
5607072 |
0.5 L | N/A | ||||||
11764498
|
Saint Gobain Life Sciences
5607077 |
10 L | N/A | ||||||
Description
- Bags are individually packaged
- Available in packs of ten
- Foil thickness: 5/10mm
- Custom sizes and configurations available on request
Specifications
Gas Sampling Bag | |
Faucet | |
Rubber |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title