missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Accessories for Digital Rockers and Waving Rotators

Product Code. 16534812 Shop All Thermo Scientific Products
Click to view available options
Description:
Dimpled Rubber Mat
Rubber Mat
Rubber Strips, 6 Each
Rubber tube replacement
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16534812

Brand: Thermo Scientific™ 88882128

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Get assorted rubberized accessories for use with Digital Rockers and Digital Waving Rotators

Assorted rubberized accessories for use with Digital Rockers and Digital Waving Rotators

Service Plan

Standard Thermo warranty guideline, provide repair service and free spare part within warranty

Warranty

2 years

TRUSTED_SUSTAINABILITY

Specifications

Product Type Rubber Tube Replacement
Material Rubber, Stainless Steel
Diameter (Metric) 44 mm
Height (Metric) 108 mm
Description Rubber tube replacement
For Use With (Equipment) Digital Vortex mixer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.