Learn More
Fisherbrand™ Rubber Pipette Bulbs for Bulb or Cylindrical Pipettes
Excellent for all media that do not attack rubber. Fisherbrand™ Pipette Bulbs come with three glassball valves, adjustable by finger pressure.
Brand:  Fisherbrand™ FB51420
Product Code. 11552672
  • £34.30 / Each
Explore more special offers
Description
Description
  • Available in a range of colors
  • With ca. 60mL volume suitable for pipettes from 2 to 50mL
  • Pipette media must not enter pipette filler; it may cause valve malfunction
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products