Learn More
Cytiva Whatman™ Rubber Stoppers
Rubber Stoppers for use with Stainless Steel Filter Funnels.
Brand: Cytiva 10446002
This item is not returnable.
View return policy
Description
- For use with Stainless Steel Filter Funnels.
Specifications
Replacement Part for GV 050 Series Glass Vacuum Filtration Manifolds | |
GV050/0/10 Stainless Steel Filter Funnels | |
Not Autoclavable | |
Rubber Stopper |
Vacuum Filtration | |
1 Ea. | |
Rubber | |
Non-sterile |