missing translation for 'onlineSavingsMsg'
Learn More

Rubidium, 99.75% (metals basis)

Product Code. p-4412893
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11371418

Brand: Thermo Scientific Alfa Aesar 010315.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-17-7 | Rb | 85.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-17-7
Molecular Formula Rb
Molecular Weight (g/mol) 85.47
MDL Number MFCD00134055
InChI Key IGLNJRXAVVLDKE-UHFFFAOYSA-N
Synonym unii-mlt4718tjw, mlt4718tjw, rubidio, rubidium-103, rubidium-106, atom, rubidium, isotope of mass 103, rubidium, isotope of mass 106, un1423 dangerous when wet
PubChem CID 5357696
ChEBI CHEBI:33322
IUPAC Name rubidium
SMILES [Rb]

Specifications

Color Silver
Appearance Silver-colored solid
Density 1.532g/cm3
Quantity 1 g
Odor Odorless
Assay Percent Range 99.75% (Metals basis)
Form Solid
Chemical Name or Material Rubidium

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.