missing translation for 'onlineSavingsMsg'
Learn More

Rubidium bromide, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11347627
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Rubidium bromide is used in infrared optics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-39-1
Molecular Formula BrRb
Molecular Weight (g/mol) 165.37
MDL Number MFCD00011186
InChI Key JAAGVIUFBAHDMA-UHFFFAOYSA-M
Synonym rubidium bromide, rubidium bromide rbbr, unii-33cm31xvqq, rubidium 1+ bromide, rubidium 1+ ion bromide, 33cm31xvqq, rubidium-bromid, rbbr, br.rb, rubidium i bromide
PubChem CID 4670918
IUPAC Name rubidium(1+);bromide
SMILES [Br-].[Rb+]

Specifications

Boiling Point 1340°C
Melting Point 693°C
Physical Form Crystalline
Quantity 5 g
Merck Index 14,8283
Solubility Information Soluble in water
Formula Weight 165.38
Percent Purity 99.8%
Odor Odorless
Refractive Index 1.553
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Rubidium bromide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.